Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dihydroxybiphenyl, 99%

Catalog No. aab2224014
Click to view available options
Quantity:
25 g
100 g
500 g

CAS: 92-88-6 | C12H10O2 | 186.21 g/mol

4,4'-Dihydroxybiphenyl is used to prepare plastics. It serves as a laboratory reagent and an intermediate in the preparation of polyether liquid crystals. Further, it is used as an antioxidant agent for rubber and latex. In addition to this, it is used in light-colored rubber products, food packaging and medical latex products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-Dihydroxybiphenyl is used to prepare plastics. It serves as a laboratory reagent and an intermediate in the preparation of polyether liquid crystals. Further, it is used as an antioxidant agent for rubber and latex. In addition to this, it is used in light-colored rubber products, food packaging and medical latex products.

Solubility
Slightly soluble in water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-88-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD00002348
InChI Key VCCBEIPGXKNHFW-UHFFFAOYSA-N
Synonym 4,4'-dihydroxybiphenyl, 4,4'-biphenol, 4,4'-biphenyldiol, biphenyl-4,4'-diol, 1,1'-biphenyl-4,4'-diol, p,p'-biphenol, p,p'-diphenol, antioxidant dod, 4,4'-dioxydiphenyl, p-dihydroxydiphenyl
PubChem CID 7112
ChEBI CHEBI:34367
IUPAC Name 4-(4-hydroxyphenyl)phenol
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)O)O

Specifications

Melting Point 279°C to 285°C
Odor Odorless
Quantity 25 g
Beilstein 1908886
Solubility Information Slightly soluble in water.
Formula Weight 186.21
Percent Purity 99%
Chemical Name or Material 4,4'-Dihydroxybiphenyl
Hazard Category H312-H315-H319-H335
Hazard Statement GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-200-5
RTECSNumber DV4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.