Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Diisocyanato-3,3'-dimethylbiphenyl 98.0+%, TCI America™
SDP

Catalog No. D2051250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2051250G 250 g
D205125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D2051250G Supplier TCI America Supplier No. D2051250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 91-97-4
Molecular Formula C16H12N2O2
Molecular Weight (g/mol) 264.284
MDL Number MFCD00019908
InChI Key ICLCCFKUSALICQ-UHFFFAOYSA-N
PubChem CID 7072
IUPAC Name 1-isocyanato-4-(4-isocyanato-3-methylphenyl)-2-methylbenzene
SMILES CC1=C(C=CC(=C1)C2=CC(=C(C=C2)N=C=O)C)N=C=O

Specifications

Melting Point 72°C
Color White-Yellow
Quantity 250 g
Synonym 3,3′-Dimethylbiphenyl-4,4′-diyl Diisocyanate
Formula Weight 264.28
Percent Purity ≥98.0% (GC)
Physical Form Pellets
Chemical Name or Material 4,4′-Diisocyanato-3,3′-dimethylbiphenyl
EINECSNumber (4)-0033
RTECSNumber DV3960000
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.