Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4'-Dimethoxybenzophenone, 98+%

Manufacturer:  Alfa Aesar™ A1146614

Quantity 25g

 View more versions of this product

Catalog No. AAA1146614


  Check Availability
Add to cart

Specifications

Description

Applications
4,4 ft.-Dimethoxybenzophenone is used in the organic synthesis. It also acts as a pharmaceutical intermediate. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
It is insoluble in water, soluble in ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible to strong oxidizing agents, strong reducing agents.
Description & Specifications

Chemical Identifiers

90-96-0
242.27
1878026
C15H14O3
MFCD00008404
4,4'-Dimethoxybenzophenone

Specifications

90-96-0
142° to 146°C
MFCD00008404
25g
≥98%
C15H14O3
242.27
1878026
4,4'-Dimethoxybenzophenone

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit