Learn More
4,4'-Dimethoxytrityl chloride, 97%
Description
4,4'-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,4′-Dimethoxytrityl chloride is used as a protective reagent for oligonucleotide synthesis. It is used for selective protection and deprotection procedures for thiol and hydroxy groups in nucleoside derivatives.
Solubility
Soluble in chloroform and methanol. Partially soluble in water
Notes
Moisture sensitive, hygroscopic. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | 121°C to 127°C |
Quantity | 100 g |
Beilstein | 2471942 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in chloroform and methanol. Partially soluble in water |
Formula Weight | 338.83 |
Percent Purity | 98% |
Chemical Name or Material | 4,4'-Dimethoxytrityl chloride |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 255-002-6 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.