Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethoxytrityl Chloride 97.0+%, TCI America™
SDP

Catalog No. D161225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D161225G 25 g
D16125G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D161225G Supplier TCI America Supplier No. D161225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 40615-36-9
Molecular Formula C21H19ClO2
Molecular Weight (g/mol) 338.83
MDL Number MFCD00008409
InChI Key JBWYRBLDOOOJEU-UHFFFAOYSA-N
Synonym 4,4'-dimethoxytrityl chloride, dmt-cl, 4,4'-dimethoxytritylchloride, 4,4'-chloro phenyl methylene bis methoxybenzene, 4,4'-dimethoxytriphenylmethyl chloride, 1,1'-chlorophenylmethylene bis 4-methoxybenzene, benzene, 1,1'-chlorophenylmethylene bis 4-methoxy, chloro-4,4'-dimethoxytriphenylmethane, 1-chloro 4-methoxyphenyl phenylmethyl-4-methoxybenzene, 4,4'-dimethoxytritylchloride dmt-cl
PubChem CID 96831
IUPAC Name 1-[chloro(4-methoxyphenyl)phenylmethyl]-4-methoxybenzene
SMILES COC1=CC=C(C=C1)C(Cl)(C1=CC=CC=C1)C1=CC=C(OC)C=C1

Specifications

Melting Point 125°C
Quantity 25 g
Formula Weight 338.83
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethoxytrityl Chloride [Hydroxyl Protecting Agent]
EINECSNumber (4)-1881
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.