Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethyl-1-pentene, 99%

Catalog No. p-4089518
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle

CAS: 762-62-9 | C7H14 | 98.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-62-9
Molecular Formula C7H14
Molecular Weight (g/mol) 98.19
MDL Number MFCD00026337
InChI Key KLCNJIQZXOQYTE-UHFFFAOYSA-N
Synonym 4,4-dimethyl-1-pentene, 1-pentene, 4,4-dimethyl, 2,2-dimethyl-4-pentene, 4,4-dimethyl-pent-1-ene, 4,4-dimethyl pent-1-ene, neoheptaene, acmc-1bcwv, ch3 3cch2ch=ch2, 4,4'-dimethyl-1-pentene
PubChem CID 12984
IUPAC Name 4,4-dimethylpent-1-ene
SMILES CC(C)(C)CC=C

Specifications

CAS Min % 98.5
CAS Max % 100.0
Density 0.6820g/mL
Boiling Point 70°C to 72°C
Flash Point -12°C
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHCH2C(CH3)3
Quantity 1 g
Specific Gravity 0.682
Formula Weight 98.19
Percent Purity 99%
Chemical Name or Material 4, 4-Dimethyl-1-pentene
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 212-101-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.