Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dimethyl-trans-stilbene 99.0+%, TCI America™
SDP

Catalog No. D270825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D270825G 25 g
D27085G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D270825G Supplier TCI America Supplier No. D270825G

Chemical Identifiers

CAS 18869-29-9
Molecular Formula C16H16
Molecular Weight (g/mol) 208.304
MDL Number MFCD00136904
InChI Key KINZBJFIDFZQCB-VAWYXSNFSA-N
PubChem CID 5462964
IUPAC Name 1-methyl-4-[(E)-2-(4-methylphenyl)ethenyl]benzene
SMILES CC1=CC=C(C=C1)C=CC2=CC=C(C=C2)C

Specifications

Melting Point 183°C
Color White
Quantity 25 g
Formula Weight 208.30
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dimethyl-trans-stilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.