Alfa Aesar™ 4,4'-Dimethylbiphenyl, 99%

Manufacturer: Alfa Aesar™ B2508914

Quantity 25g

 View more versions of this product

Catalog No. AAB2508914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 613-33-2
    Molecular Formula C14H14
    Formula Weight 182.27
    MDL Number MFCD00008545
    Beilstein 1856053
    Synonym 4,4'-Dimethyldiphenyl
    Chemical Name or Material 4,4'-Dimethylbiphenyl

    Specifications

    Formula Weight 182.27
    Molecular Formula C14H14
    MDL Number MFCD00008545
    CAS 613-33-2
    Beilstein 1856053
    Synonym 4,4'-Dimethyldiphenyl
    Boiling Point 295°C
    Melting Point 118° to 122°C
    Quantity 25g
    Chemical Name or Material 4,4'-Dimethylbiphenyl
    Assay 99%

    Description

    Applications
    4,4 ft.-Dimethylbiphenyl is used in the preparation of biphenyl-4,4’-dicarboxylic acid, which acts as a monomer utilized in the synthesis of linear long chain polymer through polycondensation reaction. It is used to prepare 4-(4 ft.-methylphenyl)benzaldehyde by selective photoxygenation reaction using 9-phenyl-10-methylacridinium perchlorate as a catalyst under visible light irradiation.

    Solubility
    Insoluble in water.

    Notes
    Incompatible with strong oxidizing agents.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit