Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4-Dimethylcyclohexanone, 97%

Catalog No. ac448810010
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle

CAS: 4255-62-3 | C8H14O | 126.199 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4255-62-3
InChI Key PXQMSTLNSHMSJB-UHFFFAOYSA-N
Synonym 4,4-dimethylcyclohexanone, cyclohexanone, 4,4-dimethyl, 4,4-dimethyl cyclohexanone, 4,4-dimethyl-cyclohexan-1-one, 4,4-dimethycyclohexanone, 4.4-dimethylcyclohexanone, acmc-209jq8, 4,4-dimethyl-cyclohexanone, ksc494a0f, 4,4-dimethyl-1-cyclohexanone
PubChem CID 138166
IUPAC Name 4,4-dimethylcyclohexan-1-one
SMILES CC1(CCC(=O)CC1)C

Specifications

Melting Point 41°C to 43°C
Boiling Point 73°C (14.0 mmHg)
Flash Point 62°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C8H14O
Linear Formula (CH3)2C6H8(=O)
Quantity 1 g
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 126.2
Formula Weight 126.2
Percent Purity 97%
Chemical Name or Material 4, 4-Dimethylcyclohexanone
Show More Show Less
Hazard Category GHS Signal Word: Danger

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.