Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dinitrodiphenyl Ether 99.0+%, TCI America™
SDP

Catalog No. D083210G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
10 g

Chemical Identifiers

CAS 101-63-3
Molecular Formula C12H8N2O5
Molecular Weight (g/mol) 260.205
MDL Number MFCD00059182
InChI Key MWAGUKZCDDRDCS-UHFFFAOYSA-N
PubChem CID 7568
IUPAC Name 1-nitro-4-(4-nitrophenoxy)benzene
SMILES C1=CC(=CC=C1[N+](=O)[O-])OC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 144°C
Color Yellow
Quantity 10 g
Synonym 4,4′-Dinitrodiphenyl Oxide
Formula Weight 260.21
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinitrodiphenyl Ether
EINECSNumber (3)-0883
RTECSNumber KN3450000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.