Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dinitrostilbene 98.0+%, TCI America™
SDP

Catalog No. D246425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D246425G 25 g
D24645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D246425G Supplier TCI America Supplier No. D246425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2501-02-2
Molecular Formula C14H10N2O4
Molecular Weight (g/mol) 270.24
MDL Number MFCD00017043
InChI Key CLVIIRIMEIEKOQ-OWOJBTEDSA-N
PubChem CID 5377860
ChEBI CHEBI:79798
IUPAC Name 1-nitro-4-[(E)-2-(4-nitrophenyl)ethenyl]benzene
SMILES C1=CC(=CC=C1C=CC2=CC=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 295°C
Color Yellow
Quantity 25 g
Formula Weight 270.24
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dinitrostilbene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.