Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4,4'-Dipyridyl disulfide, 98%

Click to view available options
Quantity:
1g
5g
2645-22-9
C10H8N2S2
220.308
MFCD00006423
UHBAPGWWRFVTFS-UHFFFAOYSA-N
4,4'-dithiodipyridine,4,4'-dipyridyl disulfide,1,2-di pyridin-4-yl disulfane,aldrithiol-4,pyridine, 4,4'-dithiobis,4,4'-dithiopyridine,di 4-pyridyl disulfide,aldrithiol 4,di 4-pyridinyl disulfide,4,4'-dipyridine disulfide
75846
CHEBI:41814
4-(pyridin-4-yldisulfanyl)pyridine
C1=CN=CC=C1SSC2=CC=NC=C2
2645-22-9
C10H8N2S2
220.308
MFCD00006423
UHBAPGWWRFVTFS-UHFFFAOYSA-N
4,4'-dithiodipyridine,4,4'-dipyridyl disulfide,1,2-di pyridin-4-yl disulfane,aldrithiol-4,pyridine, 4,4'-dithiobis,4,4'-dithiopyridine,di 4-pyridyl disulfide,aldrithiol 4,di 4-pyridinyl disulfide,4,4'-dipyridine disulfide
75846
CHEBI:41814
4-(pyridin-4-yldisulfanyl)pyridine
C1=CN=CC=C1SSC2=CC=NC=C2
Quantity 5g
Melting Point 74°C to 78°C
Beilstein 154742
Formula Weight 220.32
Percent Purity 98%
Chemical Name or Material 4,4'-Dipyridyl disulfide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C