Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Dithiodibutyric Acid 95.0+%, TCI America™
SDP

Catalog No. D175725G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 2906-60-7
Molecular Formula C8H14O4S2
Molecular Weight (g/mol) 238.316
MDL Number MFCD00004406
InChI Key YYSCJLLOWOUSHH-UHFFFAOYSA-N
PubChem CID 76196
IUPAC Name 4-(3-carboxypropyldisulfanyl)butanoic acid
SMILES C(CC(=O)O)CSSCCCC(=O)O

Specifications

Melting Point 108°C
Quantity 25 g
Synonym 4,4'-dithiodibutyric acid, butanoic acid, 4,4'-dithiobis, 4,4'-disulfanediyldibutanoic acid, 3-carboxypropyl disulfide, 4-3-carboxypropyl disulfanyl butanoic acid, 4,4-dithiodibutyric acid, butanoic acid,4,4'-dithiobis, bis 3-carboxypropyl disulfide, 4,4'-dithiodibutanoic acid, butyric acid, 4,4'-dithiodi
Formula Weight 238.32
Percent Purity ≥95.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Dithiodibutyric Acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.