Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[[(4-Fluorophenyl)imino]methyl]phenol 98.0+%, TCI America™
SDP

Catalog No. F094625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F094625G 25 g
F09465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F094625G Supplier TCI America Supplier No. F094625G

Chemical Identifiers

CAS 3382-63-6
Molecular Formula C13H10FNO
Molecular Weight (g/mol) 215.227
MDL Number MFCD00029739
InChI Key QVDWVRKNMDWNRU-UHFFFAOYSA-N
PubChem CID 5397240
IUPAC Name 4-[(4-fluoroanilino)methylidene]cyclohexa-2,5-dien-1-one
SMILES C1=CC(=O)C=CC1=CNC2=CC=C(C=C2)F

Specifications

Melting Point 181°C
Color Yellow
Quantity 25 g
Formula Weight 215.23
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-[[(4-Fluorophenyl)imino]methyl]phenol
EINECSNumber (3)-4534
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.