Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-(Hexafluoroisopropylidene)diphthalic anhydride, 99%

Catalog No. AAL1615014
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 1107-00-2 | C19H6F6O6 | 444.241 g/mol

4,4'-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4′-(Hexafluoroisopropylidene) diphthalic anhydride is involved in the synthesis of polyamides derived from terphenylenes, which is used for gas separation.

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1107-00-2
Molecular Formula C19H6F6O6
Molecular Weight (g/mol) 444.241
MDL Number MFCD00039143
InChI Key QHHKLPCQTTWFSS-UHFFFAOYSA-N
Synonym 4,4'-hexafluoroisopropylidene diphthalic anhydride, 4,4'-hexafluoroisopropylidene diphthalicanhydride, 5,5'-perfluoropropane-2,2-diyl bis isobenzofuran-1,3-dione, 2,2-bis 3,4-anhydrodicarboxyphenyl hexafluoropropane, 4,4'-hexafluoroisopropylidene bis phthalic anhydride, 6fda, 1,3-isobenzofurandione, 5,5'-2,2,2-trifluoro-1-trifluoromethyl ethylidene bis, 2,2-bis 3,4-dicarboxyphenyl-1,1,1,3,3,3-hexafluoropropane dianhydride, 5-2-1,3-dioxo-2-benzofuran-5-yl-1,1,1,3,3,3-hexafluoropropan-2-yl-2-benzofuran-1,3-dione, 2,2'-bis-3,4-dicarboxyphenyl hexafluoropropane dianhydride
PubChem CID 70677
IUPAC Name 5-[2-(1,3-dioxo-2-benzofuran-5-yl)-1,1,1,3,3,3-hexafluoropropan-2-yl]-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1C(C3=CC4=C(C=C3)C(=O)OC4=O)(C(F)(F)F)C(F)(F)F)C(=O)OC2=O

Specifications

Melting Point 244°C to 247°C
Quantity 25 g
Beilstein 7057916
Sensitivity Moisture sensitive
Solubility Information Miscible with water.
Formula Weight 444.25
Percent Purity 99%
Chemical Name or Material 4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
EINECSNumber 214-170-0
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.