Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Hexamethylenebis(1,1-dimethylsemicarbazide) 98.0+%, TCI America™
SDP

Catalog No. H106425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H106425G 25 g
H1064500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H106425G Supplier TCI America Supplier No. H106425G

Chemical Identifiers

CAS 69938-76-7
Molecular Formula C12H28N6O2
Molecular Weight (g/mol) 288.40
MDL Number MFCD00089122
InChI Key VETHREXFBVHLJJ-UHFFFAOYSA-N
Synonym 1,6-Bis[3-(dimethylamino)ureido]hexane
PubChem CID 4404942
IUPAC Name 3-(dimethylamino)-1-{6-[(N',N'-dimethylhydrazinecarbonyl)amino]hexyl}urea
SMILES CN(C)NC(=O)NCCCCCCNC(=O)NN(C)C

Specifications

Melting Point 146°C
Color White
Quantity 25 g
Formula Weight 288.40
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Hexamethylenebis(1,1-dimethylsemicarbazide)
EINECSNumber (2)-1771
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.