Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Iodophenyl)morpholine, Thermo Scientific™

Catalog No. cc17411cb
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle

Chemical Identifiers

CAS 87350-77-4
Molecular Formula C10H12INO
Molecular Weight (g/mol) 289.116
InChI Key YUEREUIIUISPFV-UHFFFAOYSA-N
PubChem CID 2795358
IUPAC Name 4-(4-iodophenyl)morpholine
SMILES C1COCCN1C2=CC=C(C=C2)I

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 4-4-iodophenyl morpholine, 4-iodophenylmorpholine, morpholine,4-4-iodophenyl, morpholine, 4-4-iodophenyl, 1-iodo-4-morpholin-4-yl benzene
Formula Weight 289.11
Percent Purity 97%
Chemical Name or Material 4-(4-Iodophenyl)morpholine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.