Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Methoxyphenyl)-2-butanone, 98%, Thermo Scientific™

Catalog No. AC313980050
Click to view available options
Quantity:
5 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-20-1
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.23
MDL Number MFCD00008791
InChI Key PCBSXBYCASFXTM-UHFFFAOYSA-N
Synonym 4-4-methoxyphenyl-2-butanone, anisylacetone, 4-4-methoxyphenyl butan-2-one, 4-methoxybenzylacetone, raspberry ketone methyl ether, 2-butanone, 4-4-methoxyphenyl, anisylacetone, p, 4-p-methoxyphenyl-2-butanone, methyl oxanone, 1-4-methoxyphenyl-3-butanone
PubChem CID 61007
IUPAC Name 4-(4-methoxyphenyl)butan-2-one
SMILES COC1=CC=C(CCC(C)=O)C=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.04g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Refractive Index 1.5190 to 1.5210 (20°C, 589nm)
Linear Formula CH3OC6H4CHCHCOCH3
Quantity 5 g
Formula Weight 178.23
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 4-(4-Methoxyphenyl)-2-butanone, 98%
Show More Show Less
EINECSNumber 203-184-2
RTECSNumber EL9085500
TSCA TSCA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.