Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[(4-Methyl-1-piperazinyl)methyl]benzoic Acid Dihydrochloride 98.0+%, TCI America™
SDP

Catalog No. M211025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M211025G 25 g
M21105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M211025G Supplier TCI America Supplier No. M211025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 106261-49-8
Molecular Formula C13H20Cl2N2O2
Molecular Weight (g/mol) 307.215
MDL Number MFCD07772867
InChI Key ISHROKOWRJDOSN-UHFFFAOYSA-N
Synonym 4-4-methylpiperazin-1-yl methyl benzoic acid dihydrochloride, 4-4-methyl-1-piperazinyl methyl benzoic acid dihydrochloride, 4-4-methylpiperazin-1-yl methyl, 4-4-methylpiperazinomethyl benzoic acid dihydrochloride, benzoic acid, 4-4-methyl-1-piperazinyl methyl-, dihydrochloride, zlchem 396, ksc909c2h, 4-4-methyl piperazino methyl benzoic acid 2hcl, 4-4-methylpiperazin-1-yl methyl benzoic acid dih
PubChem CID 10335346
IUPAC Name 4-[(4-methylpiperazin-1-yl)methyl]benzoic acid;dihydrochloride
SMILES CN1CCN(CC1)CC2=CC=C(C=C2)C(=O)O.Cl.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 307.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-[(4-Methyl-1-piperazinyl)methyl]benzoic Acid Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.