Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
1L
Packaging:
AcroSeal™ Glass Bottle
5124-30-1
C15H22N2O2
262.35
KORSJDCBLAPZEQ-UHFFFAOYSA-N
hydrogenated mdi,hylene w,bis 4-isocyanatocyclohexyl methane,4,4'-diisocyanatodicyclohexylmethane,methylene bis 4-cyclohexylisocyanate,dicyclohexylmethane 4,4'-diisocyanate,dicyclohexylmethane-4,4'-diisocyanate,methylene bis-4-cyclohexylisocyanate,nacconate h 12,cyclohexane, 1,1'-methylenebis 4-isocyanato
21202
CHEBI:53216
1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
C1CC(CCC1CC2CCC(CC2)N=C=O)N=C=O
5124-30-1
C15H22N2O2
262.35
KORSJDCBLAPZEQ-UHFFFAOYSA-N
hydrogenated mdi,hylene w,bis 4-isocyanatocyclohexyl methane,4,4'-diisocyanatodicyclohexylmethane,methylene bis 4-cyclohexylisocyanate,dicyclohexylmethane 4,4'-diisocyanate,dicyclohexylmethane-4,4'-diisocyanate,methylene bis-4-cyclohexylisocyanate,nacconate h 12,cyclohexane, 1,1'-methylenebis 4-isocyanato
21202
CHEBI:53216
1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
C1CC(CCC1CC2CCC(CC2)N=C=O)N=C=O
Boiling Point 112°C to 114°C (1010.0 hPa)
Specific Gravity 1.066
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Packaging AcroSeal™ Glass Bottle
Quantity 1L
Flash Point 113°C
Melting Point 19.0°C to 23.0°C
Density 1.0660g/mL
Color Colorless
Infrared Spectrum Authentic
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone
Formula Weight 262.35
Physical Form Liquid
Percent Purity 90%
Chemical Name or Material 4, 4'-Methylenebis(cyclohexyl isocyanate)
Show More Show Less

GHS H Statement
Fatal if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allerg

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

GHS Signal Word: Danger

EINECSNumber : 225-863-2