Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Methylenebis(cyclohexyl isocyanate), 90%, mixture of isomers, AcroSeal™

Catalog No. p-4323330
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle

CAS: 5124-30-1 | C15H22N2O2 | 262.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5124-30-1
Molecular Formula C15H22N2O2
Molecular Weight (g/mol) 262.35
MDL Number MFCD00015405
InChI Key KORSJDCBLAPZEQ-UHFFFAOYSA-N
Synonym hydrogenated mdi, hylene w, bis 4-isocyanatocyclohexyl methane, 4,4'-diisocyanatodicyclohexylmethane, methylene bis 4-cyclohexylisocyanate, dicyclohexylmethane 4,4'-diisocyanate, dicyclohexylmethane-4,4'-diisocyanate, methylene bis-4-cyclohexylisocyanate, nacconate h 12, cyclohexane, 1,1'-methylenebis 4-isocyanato
PubChem CID 21202
ChEBI CHEBI:53216
IUPAC Name 1-isocyanato-4-[(4-isocyanatocyclohexyl)methyl]cyclohexane
SMILES O=C=NC1CCC(CC2CCC(CC2)N=C=O)CC1

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 19.0°C to 23.0°C
Color Colorless
Density 1.0660g/mL
Boiling Point 112°C to 114°C (1010.0 hPa)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Quantity 100 mL
Specific Gravity 1.066
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone
Formula Weight 262.35
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 4, 4'-Methylenebis(cyclohexyl isocyanate)
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allerg
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 225-863-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.