Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Methylpiperazino)benzaldehyde, 97%, Thermo Scientific™

Catalog No. CC18704DA
Click to view available options
Quantity:
1 g

Chemical Identifiers

CAS 27913-99-1
Molecular Formula C12H17N2O
Molecular Weight (g/mol) 205.28
MDL Number MFCD00452233
InChI Key PFODEVGLOVUVHS-UHFFFAOYSA-O
Synonym 4-4-methylpiperazin-1-yl benzaldehyde, 4-4-methylpiperazino benzaldehyde, 1-4-formylphenyl-4-methylpiperazine, 4-4-methylpiperazinyl benzaldehyde, 4-4-methylpiperazino benzenecarbaldehyde, 4-4-methyl-piperazin-1-yl-benzaldehyde, benzaldehyde, 4-4-methyl-1-piperazinyl, 4-4-methyl-1-piperazinyl benzaldehyde, pubchem20530, acmc-1cazr
PubChem CID 736533
IUPAC Name 4-(4-methylpiperazin-1-yl)benzaldehyde
SMILES C[NH+]1CCN(CC1)C1=CC=C(C=O)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 4-(4-Methylpiperazino)benzaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.