Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,4'-Oxydiphthalic Anhydride 98.0+%, TCI America™
SDP

Catalog No. O0237100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0237100G 100 g
O023725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O0237100G Supplier TCI America Supplier No. O0237100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1823-59-2
Molecular Formula C16H6O7
Molecular Weight (g/mol) 310.217
MDL Number MFCD00039144
InChI Key QQGYZOYWNCKGEK-UHFFFAOYSA-N
PubChem CID 74574
IUPAC Name 5-[(1,3-dioxo-2-benzofuran-5-yl)oxy]-2-benzofuran-1,3-dione
SMILES C1=CC2=C(C=C1OC3=CC4=C(C=C3)C(=O)OC4=O)C(=O)OC2=O

Specifications

Melting Point 228°C
Quantity 100 g
Formula Weight 310.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,4′-Oxydiphthalic Anhydride
EINECSNumber (3)-3937
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.