Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Pyridyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. AAH2788203
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2788203 1 g
AAH27882MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2788203 Supplier Thermo Scientific Chemicals Supplier No. H2788203

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Self-assembly of 4-(4-pyridyl)benzoic acid (pybenH), Ni2+, SCN- and an aromatic guest (naphthalene or dibenz[a,h]anthracene) gave two isostructural inclusion compounds, in which pybenH ligands form the corresponding dimers acting as bidentate building blocks with a length of ca. 20.86 Å. By replacing an isonicotinic acid dimer with a 4-(4-pyridyl)benzoic acid (pybenH) dimer, new inclusion compounds in which pybenH dimers function as bridging ligands of ca. 21 Å.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4385-76-6
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD04114574
InChI Key DZLGZIGLHCRIMF-UHFFFAOYSA-N
Synonym 4-pyridin-4-yl-benzoic acid, 4-4-pyridyl benzoic acid, 4-pyridin-4-yl benzoic acid, 4-pyrid-4-ylbenzoic acid, 4-4-pyridinyl benzoic acid, 4-4-carboxyphenyl pyridine, 4'-pyridyl-4-benzoic acid, benzoic acid, 4-4-pyridinyl, pubchem22345, acmc-1adxv
PubChem CID 1520811
IUPAC Name 4-(pyridin-4-yl)benzoic acid
SMILES OC(=O)C1=CC=C(C=C1)C1=CC=NC=C1

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 199.2
Percent Purity 97%
Chemical Name or Material 4-(4-Pyridyl)benzoic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.