Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4',5,7-Trihydroxyflavone, 97%

Catalog No. aal15041mb
Click to view available options
Quantity:
25 mg
100 mg

CAS: 520-36-5 | C15H10O5 | 270.24 g/mol

4',5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′,5,7-Trihydroxyflavone is a nonmutagenic flavonoid shown to inhibit cell proliferation, angiogenesis and protein kinase. Also induces apoptosis in breast cancer cells. It inhibit topoisomerase I-catalyzed DNA re-ligation and enhance gap junctional intercellular communication.

Solubility
Soluble in dimethyl sulfoxide (27 mg/ml) or 1M potassium hydroxide (50mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 520-36-5
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00006831
InChI Key KZNIFHPLKGYRTM-UHFFFAOYSA-N
Synonym apigenin, 5,7-dihydroxy-2-4-hydroxyphenyl-4h-chromen-4-one, chamomile, spigenin, versulin, apigenol, 4',5,7-trihydroxyflavone, apigenine, c.i. natural yellow 1, 5,7,4'-trihydroxyflavone
PubChem CID 5280443
ChEBI CHEBI:18388
IUPAC Name 5,7-dihydroxy-2-(4-hydroxyphenyl)chromen-4-one
SMILES OC1=CC=C(C=C1)C1=CC(=O)C2=C(O)C=C(O)C=C2O1

Specifications

Melting Point ∽315°C (decomposition)
Quantity 25 mg
Beilstein 262620
Merck Index 14,730
Solubility Information Soluble in dimethyl sulfoxide (27mg/ml) or 1M potassium hydroxide (50mg/ml).
Formula Weight 270.24
Percent Purity 97%
Chemical Name or Material 4',5,7-Trihydroxyflavone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-292-3
RTECSNumber LK9276000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.