Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine 99.0+%, TCI America™
SDP

Catalog No. C0932100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0932100MG 100 mg
C09321G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0932100MG Supplier TCI America Supplier No. C0932100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33006-91-6
Molecular Formula C11H10ClN5
Molecular Weight (g/mol) 247.686
MDL Number MFCD00059781
InChI Key WLNTVTDTBKPTCA-UHFFFAOYSA-N
PubChem CID 118377
IUPAC Name 4-[(5-chloropyridin-2-yl)diazenyl]benzene-1,3-diamine
SMILES C1=CC(=C(C=C1N)N)N=NC2=NC=C(C=C2)Cl

Specifications

Quantity 100 mg
Formula Weight 247.69
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(5-Chloro-2-pyridylazo)-1,3-phenylenediamine [for Colorimetric Analysis of Co, Cd]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.