Learn More
4,5-Diazafluoren-9-one, 98%
Description
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,5-Diazafluoren-9-one behaves like cyclopentadienones and reacts with [Cp*Co(C2H4)2] to form air -stable sandwich-type complexes [Cp*Co(η4-2)].
Solubility
Soluble in dichloromethane, tetrahydrofuran, chloroform, benzene, toluene and polar organic solvents. Insoluble in diethyl ether and alkanes.
Notes
Incompatible with strong oxidizing agents. Avoid heat and source of ignition.

Specifications
Specifications
Melting Point | 214°C to 217°C |
Quantity | 5 g |
Solubility Information | Soluble in dichloromethane,tetrahydrofuran,chloroform,benzene,toluene and polar organic solvents. Insoluble in diethyl ether and alkanes. |
Formula Weight | 182.18 |
Percent Purity | 98% |
Chemical Name or Material | 4,5-Diazafluoren-9-one |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.