Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dibromo-1,2-phenylenediamine 98.0+%, TCI America™
SDP

Catalog No. D406225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D406225G 25 g
D40625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D406225G Supplier TCI America Supplier No. D406225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49764-63-8
Molecular Formula C6H6Br2N2
Molecular Weight (g/mol) 265.94
MDL Number MFCD00795598
InChI Key TTXGKCVKGXHPRI-UHFFFAOYSA-N
Synonym 1,2-Diamino-4,5-dibromobenzene
PubChem CID 12196944
IUPAC Name 4,5-dibromobenzene-1,2-diamine
SMILES NC1=CC(Br)=C(Br)C=C1N

Specifications

Melting Point 137°C
Quantity 25 g
Formula Weight 265.94
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4,5-Dibromo-1,2-phenylenediamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.