Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4',5'-Dichloro-2'-nitroacetanilide 97.0+%, TCI America™
SDP

Catalog No. D490725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D490725G 25 g
D49075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D490725G Supplier TCI America Supplier No. D490725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5462-30-6
Molecular Formula C8H6Cl2N2O3
Molecular Weight (g/mol) 249.05
MDL Number MFCD00024301
InChI Key ZEGRPTYRAGSSBH-UHFFFAOYSA-N
Synonym n-4,5-dichloro-2-nitrophenyl acetamide, n1-4,5-dichloro-2-nitrophenyl acetamide, acetamide, n-4,5-dichloro-2-nitrophenyl, 4',5'-dichloro-2'-nitroacetanilide, 4,5-dichloro-2-nitroacetanilide, acmc-20akbu, pubchem22538, ksc496o3f, 4,5-dichloro-2-nitro-n-acetylaniline, acetamide,n-4,5-dichloro-2-aophenyl
PubChem CID 226498
IUPAC Name N-(4,5-dichloro-2-nitrophenyl)acetamide
SMILES CC(=O)NC1=CC(Cl)=C(Cl)C=C1[N+]([O-])=O

Specifications

Melting Point 126°C
Quantity 25 g
Formula Weight 249.05
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′,5′-Dichloro-2′-nitroacetanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.