Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dicyanoimidazole, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
1122-28-7
C5H2N4
118.1
MFCD00005194
XGDRLCRGKUCBQL-UHFFFAOYSA-N
4,5-dicyanoimidazole,imidazole-4,5-dicarbonitrile,4,5-imidazoledicarbonitrile,activator,pubchem7602,4, 5-dicyanoimidazole,moa 7,imidazole-4,5-dinitrile,acmc-1brr7,4,5-dicyano-1h-imidazole
70729
1H-imidazole-4,5-dicarbonitrile
C1=NC(=C(N1)C#N)C#N
1122-28-7
C5H2N4
118.1
MFCD00005194
XGDRLCRGKUCBQL-UHFFFAOYSA-N
4,5-dicyanoimidazole,imidazole-4,5-dicarbonitrile,4,5-imidazoledicarbonitrile,activator,pubchem7602,4, 5-dicyanoimidazole,moa 7,imidazole-4,5-dinitrile,acmc-1brr7,4,5-dicyano-1h-imidazole
70729
1H-imidazole-4,5-dicarbonitrile
C1=NC(=C(N1)C#N)C#N
Melting Point 174.0°C to 177.0°C
Quantity 25g
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (HPLC)
Formula Weight 118.1
Physical Form Crystalline Powder
Percent Purity 99%
Chemical Name or Material 4, 5-Dicyanoimidazole, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Immediately call a POISON CENTER or

GHS Signal Word: Danger

EINECSNumber : 214-344-6