Learn More
4,5-Dimethoxy-o-phenylenediamine dihydrochloride, 98%
Description
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2'-dithiobis(1-aminonaphthalene).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4,5-Dimethoxy-o-phenylenediamine dihydrochloride is used to prepare highly fluorescent benzimidazole derivatives by reacting with aldehydes. It is used for the detection of aromatic aldehydes and 2,2′-dithiobis(1-aminonaphthalene).
Solubility
Soluble in warm methanol, dimethyl sulfoxide and water.
Notes
Incompatible with oxidizing agents.

Specifications
Specifications
Melting Point | ∽265°C (decomposition) |
Quantity | 250 mg |
UN Number | UN2811 |
Beilstein | 3714004 |
Solubility Information | Soluble in warm methanol,dimethyl sulfoxide and water. |
IUPAC Name | 4,5-dimethoxybenzene-1,2-diamine;dihydrochloride |
Formula Weight | 241.12 |
Percent Purity | 98% |
Chemical Name or Material | 4,5-Dimethoxy-o-phenylenediamine dihydrochloride |
Safety and Handling
Hazard Category | H302+H312+H332-H315-H319-H335 |
Hazard Statement | GHS H Statement H331-H302-H312-H315-H319-H335 Toxic if inhaled. Harmful if swallowed. Harmful in contact with skin. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.