Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethylthiazole, 98%

Catalog No. aaa1078506
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 3581-91-7 | C5H7NS | 113.178 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3581-91-7
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00005336
InChI Key UWSONZCNXUSTKW-UHFFFAOYSA-N
Synonym 4,5-dimethylthiazole, thiazole, 4,5-dimethyl, 4,5-dimethyl thiazole, 4,5-dimethyl-thiazole, unii-u3rp5i088g, fema no. 3274, pubchem9938, acmc-209ijb, dimethylthiazole, 4,5, 4-27-00-00986 beilstein handbook reference
PubChem CID 62510
IUPAC Name 4,5-dimethyl-1,3-thiazole
SMILES CC1=C(SC=N1)C

Specifications

Density 1.07
Boiling Point 157°C to 158°C
Flash Point 51°C (123°F)
Refractive Index 1.521
Quantity 5 g
UN Number UN1993
Beilstein 105694
Formula Weight 113.18
Percent Purity 98%
Chemical Name or Material 4,5-Dimethylthiazole
Hazard Category H226-H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H226
Toxic if swallowed.
Flammable liquid and vapor.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 222-703-3
RTECSNumber XJ4380000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.