Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,5-Dimethylthiophene-2-carboxaldehyde, 97%, Thermo Scientific™

Catalog No. AC342280010
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 5928-48-3
Molecular Formula C7H8OS
Molecular Weight (g/mol) 140.2
MDL Number MFCD02093897
InChI Key QSBBXKVGLJSGAJ-UHFFFAOYSA-N
Synonym 4,5-dimethylthiophene-2-carboxaldehyde, 2-thiophenecarboxaldehyde, 4,5-dimethyl, acmc-20amru, pubchem20559, 2,3-dimethyl-5-formylthiophene, 2-thiophenecarboxaldehyde,4,5-dimethyl, 4,5-dimethyl-2-thiophene carboxaldehyde
PubChem CID 2733926
IUPAC Name 4,5-dimethylthiophene-2-carbaldehyde
SMILES CC1=C(SC(=C1)C=O)C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow to Brown
Boiling Point 80°C to 85°C (0.2 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5780 to 1.58
Quantity 1 g
Beilstein 17, IV, 4554
Formula Weight 140.2
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 4, 5-Dimethylthiophene-2-carboxaldehyde, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Product darkens on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.