Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Bis(diphenylphosphino)phenoxazine, 98+%

Catalog No. AAH2733003
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2733003 1 g
AAH27330MD 250 mg
AAH2733006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2733003 Supplier Thermo Scientific Chemicals Supplier No. H2733003
Add to Cart

CAS: 261733-18-0 | C36H27NOP2 | 551.57 g/mol

4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Bis(diphenylphosphino)phenoxazine used as an intermediate in the manufacture of pharmaceuticals, agrochemicals, dyestuffs, fine chemicals, medical and material synthesis.

Solubility
Miscible with water.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261733-18-0
Molecular Formula C36H27NOP2
Molecular Weight (g/mol) 551.57
MDL Number MFCD03788937
InChI Key HSWZLYXRAOXOLL-UHFFFAOYSA-N
Synonym 4,6-bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-10h-phenoxazine, nixantphos, 10h-phenoxazine, 4,6-bis diphenylphosphino, 4,6-bis diphenylphosphanyl-10h-phenoxazine, nixanthphos, n-xantphos, acmc-209gp1, bis diphenylphosphino phenoxazine, 4,6-bis diphenylphosphino-phenoxazine
PubChem CID 3255346
IUPAC Name (6-diphenylphosphanyl-10H-phenoxazin-4-yl)-diphenylphosphane
SMILES N1C2=C(OC3=C1C=CC=C3P(C1=CC=CC=C1)C1=CC=CC=C1)C(=CC=C2)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 251°C to 254°C
Quantity 1 g
Solubility Information Miscible with water.
Formula Weight 551.55
Percent Purity ≥98%
Chemical Name or Material 4,6-Bis(diphenylphosphino)phenoxazine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.