Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Di-tert-butyl-m-cresol 96.0+%, TCI America™
SDP

Catalog No. d143225g Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g

Chemical Identifiers

CAS 497-39-2
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
MDL Number MFCD00059208
InChI Key WYSSJDOPILWQDC-UHFFFAOYSA-N
Synonym 2,4-Di-tert-butyl-5-hydroxytoluene, 2,4-Di-tert-butyl-5-methylphenol
PubChem CID 10346
IUPAC Name 2,4-ditert-butyl-5-methylphenol
SMILES CC1=CC(=C(C=C1C(C)(C)C)C(C)(C)C)O

Specifications

Melting Point 62°C
Boiling Point 282°C
Quantity 25 g
Formula Weight 220.36
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Di-tert-butyl-m-cresol
EINECSNumber (3)-0540
RTECSNumber GO7800000
TSCA Yes
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.