Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dichloro-5-methoxypyrimidine 98.0+%, TCI America™
SDP

Catalog No. D472325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D472325G 25 g
D47235G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D472325G Supplier TCI America Supplier No. D472325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5018-38-2
Molecular Formula C5H4Cl2N2O
Molecular Weight (g/mol) 179
MDL Number MFCD00087680
InChI Key IJQIGKLDBGKSNT-UHFFFAOYSA-N
Synonym 4,6-dichlor-5-methoxypyrimidin, 4,6-dichloro-5-methoxy-pyrimidine, 4,6-dichloro-5-methoxy pyrimidine, zlchem 448, pubchem7077, pyrimidine, 4,6-dichloro-5-methoxy, acmc-1avm3, 4,6 dichloro-5-methoxypyrimidine, pyrimidine,6-dichloro-5-methoxy, 4,6-dichloro-5-methoxypyrimidine
PubChem CID 78720
IUPAC Name 4,6-dichloro-5-methoxypyrimidine
SMILES COC1=C(N=CN=C1Cl)Cl

Specifications

Melting Point 57°C
Color Yellow
Quantity 25 g
Formula Weight 179.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloro-5-methoxypyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.