Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dichloropyrimidine 98.0+%, TCI America™
SDP

Catalog No. D248825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D248825G 25 g
D24885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D248825G Supplier TCI America Supplier No. D248825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1193-21-1
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00006109
InChI Key XJPZKYIHCLDXST-UHFFFAOYSA-N
Synonym pyrimidine, 4,6-dichloro, 4,6-dichloro-pyrimidine, 4,6-dichloro pyrimidine, 4,6-dichloro-1,3-diazine, 4,6-dichlorpyrimidin, dichloropyrmidine, dichloropyrimidine, zlchem 225, pubchem6913, 4,6-dichloropyrimdine
PubChem CID 70943
IUPAC Name 4,6-dichloropyrimidine
SMILES ClC1=CC(Cl)=NC=N1

Specifications

Melting Point 68°C
Color White-Yellow
Quantity 25 g
UN Number 3263
Formula Weight 148.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dichloropyrimidine
EINECSNumber (5)-0927
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.