Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98+%

Catalog No. AAL0716006
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0716006 5 g
AAL07160MD 250 mg
AAL0716003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0716006 Supplier Thermo Scientific Chemicals Supplier No. L0716006
Add to Cart

CAS: 2465-59-0 | C5H4N4O2 | 152.113 g/mol

4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine is an inhibitor to study the specificity and kinetics of xanthine oxidase. It is also used as an anti-inflammatory agent via inhibition of xanthine oxidase.

Solubility
Soluble in DMSO.

Notes
Incompatible materials are Strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2465-59-0
Molecular Formula C5H4N4O2
Molecular Weight (g/mol) 152.113
MDL Number MFCD00056934
InChI Key HXNFUBHNUDHIGC-UHFFFAOYSA-N
Synonym oxypurinol, oxipurinol, alloxanthine, 1h-pyrazolo 3,4-d pyrimidine-4,6-diol, oxoallopurinol, 4,6-dihydroxypyrazolo 3,4-d pyrimidine, 1h-pyrazolo 3,4-d pyrimidine-4,6 5h,7h-dione, oxipurinolum, 1h-pyrazolo 3,4-d pyrimidine-4,6 2h,5h-dione, alloxanthin van
PubChem CID 4644
ChEBI CHEBI:28315
IUPAC Name 1,2-dihydropyrazolo[3,4-d]pyrimidine-4,6-dione
SMILES C1=C2C(=NC(=O)NC2=O)NN1

Specifications

Melting Point >300°C
Odor Odorless
Quantity 5 g
Beilstein 139956
Solubility Information Soluble in DMSO.
Formula Weight 152.11
Percent Purity ≥98%
Chemical Name or Material 4,6-Dihydroxy-1H-pyrazolo[3,4-d]pyrimidine
Hazard Category H301-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 219-570-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.