Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dimethyl-2-mercaptopyrimidine 98.0+%, TCI America™
SDP

Catalog No. D1580250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1580250G 250 g
D158025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1580250G Supplier TCI America Supplier No. D1580250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22325-27-5
Molecular Formula C6H8N2S
Molecular Weight (g/mol) 140.204
MDL Number MFCD00006079
InChI Key RAFAYWADRVMWFA-UHFFFAOYSA-N
Synonym 4,6-dimethyl-2-mercaptopyrimidine, 4,6-dimethyl-2 1h-pyrimidinethione, 4,6-dimethylpyrimidine-2-thiol, 4,6-dimethyl-2-thiopyrimidine, 4,6-dimethyl-2-pyrimidinethiol, 2-mercapto-4,6-dimethylpyrimidine, 2 1h-pyrimidinethione, 4,6-dimethyl, 4,6-dimethylpyrimidine-2-thione, 4,6-dimethyl-pyrimidine-2-thiol, 4,6-dimethyl-2-thiolpyrimidine
PubChem CID 673664
IUPAC Name 4,6-dimethyl-1H-pyrimidine-2-thione
SMILES CC1=CC(=NC(=S)N1)C

Specifications

Melting Point 219°C
Color Yellow
Quantity 250 g
Formula Weight 140.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,6-Dimethyl-2-mercaptopyrimidine
EINECSNumber (5)-0922
RTECSNumber UW5650000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.