Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,6-Dimethyldibenzothiophene, 97%

Catalog No. p-7036971
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 1207-12-1 | C14H12S | 212.31 g/mol

4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,6-Dimethyldibenzothiophene is used in petroleum desulfurization studies.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1207-12-1
Molecular Formula C14H12S
Molecular Weight (g/mol) 212.31
MDL Number MFCD00216264
InChI Key KMPJENUWHPZRGZ-UHFFFAOYSA-N
Synonym dibenzothiophene, 4,6-dimethyl, 4,6-dimethyldibenzo b,d thiophene, 4,6-dmdbt, 6,10-dimethyl-8-thiatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene, 4,6-dimethyl-dibenzothiophen, ksc491i3n, bidd:gt0307, 4,6-dimethyl dibenzo b thiophene, 4,6-dimethyldibenzothiophene, 4,6-dimethylbenzo b benzo b thiophene
PubChem CID 1268103
IUPAC Name 4,6-dimethyldibenzothiophene
SMILES CC1=CC2=C(SC3=C2C(C)=CC=C3)C=C1

Specifications

Melting Point 153°C to 157°C
Quantity 250 mg
Formula Weight 212.31
Percent Purity 97%
Chemical Name or Material 4,6-Dimethyldibenzothiophene
Hazard Category H302
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 214-894-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.