Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(6-Isocyanatopyrid-2-yl)morpholine, Tech., Thermo Scientific™

Catalog No. CC38306DA
Click to view available options
:
Amber Glass Bottle; 1g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 884507-15-7
CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Molecular Formula C10H11N3O2
Quantity 1g
MDL Number MFCD08435861
Synonym 4-6-isocyanatopyrid-2-yl morpholine, 4-6-isocyanatopyridin-2-yl morpholine, 6-morpholin-4-ylpyridin-2-isocyanate, morpholine,4-6-isocyanato-2-pyridinyl
InChI Key RYYZXDNHAQDJTI-UHFFFAOYSA-N
SMILES C1COCCN1C2=CC=CC(=N2)N=C=O
IUPAC Name 4-(6-isocyanatopyridin-2-yl)morpholine
Molecular Weight (g/mol) 205.217
PubChem CID 18525736
Formula Weight 205.22
Percent Purity Tech.
Grade Technical
Chemical Name or Material 4-(6-Isocyanatopyrid-2-yl)morpholine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.