Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dibromo-1,10-phenanthroline Hydrate 98.0+%, TCI America™
SDP

Catalog No. D49871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D49871G 1 g
D4987200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D49871G Supplier TCI America Supplier No. D49871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 156492-30-7
Molecular Formula C12H6Br2N2
Molecular Weight (g/mol) 338.002
MDL Number MFCD25562940
InChI Key AKZAIDYHEKUXBU-UHFFFAOYSA-N
PubChem CID 11393583
IUPAC Name 4,7-dibromo-1,10-phenanthroline
SMILES C1=CC2=C(C=CN=C2C3=NC=CC(=C31)Br)Br

Specifications

Melting Point 236°C
Color Yellow
Quantity 1 g
Formula Weight 338.00
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dibromo-1,10-phenanthroline Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.