Alfa Aesar™ 4',7-Dihydroxyisoflavone, 97%

Manufacturer: Alfa Aesar™ B2287714

Quantity 25g

 View more versions of this product

Catalog No. AAB2287714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 486-66-8
    Molecular Formula C15H10O4
    Formula Weight 254.24
    MDL Number MFCD00016954
    Beilstein 231523
    Merck Index 14,2801
    Synonym Daidzein
    Chemical Name or Material 4',7-Dihydroxyisoflavone

    Specifications

    Formula Weight 254.24
    MDL Number MFCD00016954
    Merck Index 14,2801
    Melting Point 328° to 332°C (dec.)
    Quantity 25g
    Molecular Formula C15H10O4
    CAS 486-66-8
    Beilstein 231523
    Synonym Daidzein
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number DJ3100040
    Chemical Name or Material 4',7-Dihydroxyisoflavone
    Assay 97%

    Description

    Applications
    4 ft.,7-Dihydroxyisoflavone is a phytoestrogen that is suggested to play a role in preventing hormone-induced cancers. It protects against oxidative damage in liver cells induced by 7,12-dimethylbenz[a]anthracene (DMBA). Catalase and superoxide dismutase activity, down-regulated by DMBA, was restored by daidzein.

    Solubility
    Insoluble in water. Soluble in DMSO (25 mg/ml), ethanol (2 mg/ml), DMF (10 mg/ml), and 1:10 DMSO:PBS (pH 7.2) (0.15 mg/ml).

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit