Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,7-Dimethoxy-1,10-phenanthroline, 97%

Catalog No. p-4228612
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle

CAS: 92149-07-0 | C14H12N2O2 | 240.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92149-07-0
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00233883
InChI Key ZPGVCQYKXIQWTP-UHFFFAOYSA-N
Synonym 4,7-dimethoxy-1,10-phenanthroline, acmc-20a0a1, 4,7-dimethoxy-1,10-phenthroline, 4?7-dimethoxy-1?10-phenanthroline, 1,10-phenanthroline, 4,7-dimethoxy, 4,7-bis methoxy-1,10-phenanthroline, 4,7-bis methyloxy-1,10-phenanthroline
PubChem CID 10462298
IUPAC Name 4,7-dimethoxy-1,10-phenanthroline
SMILES COC1=C2C=CC3=C(C=CN=C3C2=NC=C1)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 212.0°C to 214.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 250 mg
Formula Weight 240.26
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 4,7-Dimethoxy-1,10-phenanthroline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Causes serious eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.