Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene 95.0+%, TCI America™
SDP

Catalog No. B4395200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4395200MG 200 mg
B43951G 1 g
B43955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B4395200MG Supplier TCI America Supplier No. B4395200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1160823-77-7
Molecular Formula C26H38O2S2
Molecular Weight (g/mol) 446.708
MDL Number MFCD17018551
InChI Key HATOWNJGYIVNBU-UHFFFAOYSA-N
PubChem CID 53486668
IUPAC Name 4,8-bis(2-ethylhexoxy)thieno[2,3-f][1]benzothiole
SMILES CCCCC(CC)COC1=C2C=CSC2=C(C3=C1SC=C3)OCC(CC)CCCC

Specifications

Quantity 200 mg
Formula Weight 446.71
Percent Purity ≥95.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 4,8-Bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b′]dithiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.