Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidobenzoic acid, 99+%

Catalog No. p-47711
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle

CAS: 556-08-1 | C9H9NO3 | 179.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 556-08-1
Molecular Formula C9H9NO3
Molecular Weight (g/mol) 179.17
MDL Number MFCD00002534
InChI Key QCXJEYYXVJIFCE-UHFFFAOYSA-N
Synonym acedoben, p-acetamidobenzoic acid, 4-acetylamino benzoic acid, 4-carboxyacetanilide, 4-acetylaminobenzoic acid, benzoic acid, 4-acetylamino, p-acetylamino benzoic acid, p-acetoaminobenzoic acid, p-acetaminobenzoic acid, n-acetyl-p-aminobenzoic acid
PubChem CID 19266
ChEBI CHEBI:46171
IUPAC Name 4-acetamidobenzoic acid
SMILES CC(=O)NC1=CC=C(C=C1)C(=O)O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 255°C to 262°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Plastic bottle
Linear Formula CH3CONHC6H4CO2H
Quantity 25 g
Beilstein 14, 432
Formula Weight 179.17
Percent Purity 99+%
Physical Form Powder
Chemical Name or Material 4-Acetamidobenzoic acid, 99+%
Show More Show Less
EINECSNumber 209-114-7
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.