Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetamidopiperidine 98.0+%, TCI America™
SDP

Catalog No. A165325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A165325G 25 g
A16535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A165325G Supplier TCI America Supplier No. A165325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5810-56-0
Molecular Formula C7H14N2O
Molecular Weight (g/mol) 142.202
MDL Number MFCD05254805
InChI Key YLWUSMHZABTZGP-UHFFFAOYSA-N
Synonym 4-acetamidopiperidine, n-piperidin-4-yl acetamide, 4-acetylamino-piperidine, 4-acetylaminopiperidine, n-piperidin-4-yl-acetamide, acetamide, n-4-piperidinyl, 4-acetylamino piperidine, n-4-piperidinyl acetamide, acmc-20aiud, pubchem9227
PubChem CID 1445156
IUPAC Name N-piperidin-4-ylacetamide
SMILES CC(=O)NC1CCNCC1

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 25 g
Formula Weight 142.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Acetamidopiperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.