Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
4'-Acetoxyacetophenone, 99%
Click to view available options
Quantity:
25 g
100 g
Description
4'-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4′-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.
Solubility
Slightly soluble in water(4450 mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
4′-Acetoxyacetophenone it can react with ethane-1,2-diol to get 2-(4-acetoxyphenyl)-2-methyl-1,3-dioxolane with yield is 67 %.
Solubility
Slightly soluble in water(4450 mg/L).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Specifications
Specifications
Melting Point | 51°C to 54°C |
Boiling Point | 165°C to 170°C (14 mmHg) |
Quantity | 25 g |
Beilstein | 2093080 |
Solubility Information | Slightly soluble in water(4450mg/L). |
Formula Weight | 178.19 |
Percent Purity | 99% |
Chemical Name or Material | 4'-Acetoxyacetophenone |
Safety and Handling
EINECSNumber | 235-894-3 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction