Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetoxystyrene, 95%, stab.

Catalog No. p-7045687
Click to view available options
Quantity:
2 g
10 g

CAS: 2628-16-2 | C10H10O2 | 162.188 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2628-16-2
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.188
MDL Number MFCD00075734
InChI Key JAMNSIXSLVPNLC-UHFFFAOYSA-N
Synonym 4-acetoxystyrene, 4-vinylphenyl acetate, 4-ethenylphenol acetate, p-acetoxystyrene, phenol, 4-ethenyl-, acetate, p-vinylphenol acetate, unii-7s21904ayn, phenol, 4-ethenyl-, 1-acetate, 4-ethenylphenyl acetate, acetic acid 4-vinylphenyl ester
PubChem CID 75821
IUPAC Name (4-ethenylphenyl) acetate
SMILES CC(=O)OC1=CC=C(C=C1)C=C

Specifications

Melting Point 7°C to 8°C
Density 1.06
Boiling Point 259°C to 260°C
Flash Point 87°C (188°F)
Refractive Index 1.538
Quantity 2 g
Beilstein 1862793
Formula Weight 162.19
Percent Purity 95%
Chemical Name or Material 4-Acetoxystyrene, stabilized
Hazard Category H227-H302-H315-H317-H319-H334
Hazard Statement GHS H Statement
H334-H302-H227-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P210-P261-P264b-P270-P272-P280-P285-P301+P312-P302+P352-P304+P341-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
EINECSNumber 434-600-2
RTECSNumber SL3784000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.