Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Acetylbenzenesulfonamide, 97%

Catalog No. p-7031727
Click to view available options
Quantity:
1 g
5 g

CAS: 1565-17-9 | C8H9NO3S | 199.22 g/mol

4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-acetylbenzenesulfonamide and acetohexamide which is used to characterize the pharmacokinetics of a reversible drug—metabolite system.

Solubility
soluble in water( 2650 mg/L).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1565-17-9
Molecular Formula C8H9NO3S
Molecular Weight (g/mol) 199.22
MDL Number MFCD00792524
InChI Key CSATVXJBGFVJES-UHFFFAOYSA-N
Synonym 4-acetylbenzenesulphonamide, p-acetylbenzenesulfonamide, 4-acetyl-benzenesulfonamide, 4-acetylbenzene-1-sulfonamide, benzenesulfonamide, 4-acetyl, p-acetobenzenesulfonamide, benzenesulfonamide, p-acetyl-6ci,7ci,8ci, 4-acetyl-benzenesulfonamid, p-acetylbenzene sulfonamide, #
PubChem CID 74065
IUPAC Name 4-acetylbenzenesulfonamide
SMILES CC(=O)C1=CC=C(C=C1)S(N)(=O)=O

Specifications

Melting Point 177°C to 179°C
Quantity 1 g
Solubility Information soluble in water( 2650mg/L).
Formula Weight 199.23
Percent Purity 97%
Chemical Name or Material 4-Acetylbenzenesulfonamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 216-363-5
RTECSNumber DA9386000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.